missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ G3BP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553797
This item is not returnable.
View return policy
Description
Immunogen sequence: KNLEELEEKS TTPPPAEPVS LPQEPPKPRV EAKPEVQSQP PRVREQRPRE RPGFPPRGPR PGRGDMEQND S Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.
G3BP2 gene ontology annotations related to this gene include stress granule assembly.
Specifications
| G3BP2 | |
| Polyclonal | |
| Unconjugated | |
| G3BP2 | |
| AA409541; E430034L04Rik; G3BP; G3BP stress granule assembly factor 2; G3bp2; G3BP-2; GAP SH3 domain-binding protein 2; GTPase activating protein (SH3 domain) binding protein 2; KIAA0660; mKIAA0660; Ras GTPase-activating protein-binding protein 2; Ras-GTPase activating protein SH3 domain-binding protein 2; RGD1309571 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9908 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UN86 | |
| G3BP2 | |
| Recombinant protein corresponding to Human G3BP2. Recombinant protein control fragment (Product #RP-91850). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering