missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAB2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97204AF405
This item is not returnable.
View return policy
Description
GAB2 Polyclonal antibody specifically detects GAB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GAB2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| GRB2-associated binding protein 2, GRB2-associated-binding protein 2, Growth factor receptor bound protein 2-associated protein 2, KIAA0571GRB2-associated binder 2, pp100 | |
| Produced in rabbits immunized with E. coli-derived Human GAB2 fragment. | |
| 0.1 mL | |
| Cancer | |
| 9846 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction