missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R gamma 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB300-151
This item is not returnable.
View return policy
Description
GABA-A R gamma 2 Polyclonal specifically detects GABA-A R gamma 2 in Human, Mouse, Rat, Turkey, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.
Specifications
| GABA-A R gamma 2 | |
| Polyclonal | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen | |
| P22723, P18508 | |
| GABRG2 | |
| Synthetic peptide corresponding to amino acid residues specific to the gamma 2 subunit conjugated to KLH. Accession # P18508 | |
| Unpurified | |
| RUO | |
| Primary | |
| Specific for the ∼46k gamma2-subunit of the GABAA receptor in Western blots of rat brain extracts. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Neat whole antisera with No Preservative | |
| CAE2, ECA2, GABA(A) receptor subunit gamma-2, gamma-aminobutyric acid (GABA) A receptor, gamma 2, gamma-aminobutyric acid receptor subunit gamma-2, GEFSP3 | |
| Rabbit | |
| 46 kDa | |
| 0.05 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2566 | |
| Human, Mouse, Rat, Turkey, Zebrafish | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction