missing translation for 'onlineSavingsMsg'
Learn More

GABA-AR alpha 4 Antibody (S398A-34), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-59326-25ug

 View more versions of this product

Product Code. 18671578

  • € 174.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GABA-AR alpha 4 Monoclonal specifically detects GABA-AR alpha 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GABA-AR alpha 4
Monoclonal
Unconjugated
PBS (pH 7.4), 50% glycerol with 0.1% Sodium Azide
GABRA4
Fusion protein amino acids 410-450 (Cytoplasmic C-terminus) of rat GABA-A-Receptor Alpha 4
25 μg
Neuronal Cell Markers, Neuroscience, Neurotransmission
2557
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
S398A-34
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
GABA(A) receptor subunit alpha-4, gamma-aminobutyric acid (GABA) A receptor, alpha 4, gamma-aminobutyric acid receptor subunit alpha-4
Mouse
Protein G purified
RUO
Primary
Detects 60kDa. Does not cross-react with other GABA-A-Receptor Alpha subunits.
Store at -20°C.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only