missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 zeta, p Tyr72 Antibody (EM-26), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62193
This item is not returnable.
View return policy
Description
CD3 zeta Monoclonal antibody specifically detects CD3 zeta in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular), Immunocytochemistry
Specifications
| CD3 zeta | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| EM-26 | |
| Western Blot 2-5 μg/mL, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular) 1-9 μg/mL, Immunocytochemistry | |
| CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA | |
| A phospho specific peptide corresponding to the amino acids surrounding tyrosine 72 of mouse CD3 zeta linked to KLH | |
| 0.1 mg | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 919 | |
| Store at 4°C. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction