missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 5 Antibody (OTI1G9), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70789UV
This item is not returnable.
View return policy
Description
GABA-AR alpha 5 Monoclonal specifically detects GABA-AR alpha 5 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GABA-AR alpha 5 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GABRA5 | |
| Recombinant protein fragment corresponding to amino acids 32-259 of human GABA-A R alpha 5 (NP_000801) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot | |
| OTI1G9 | |
| Western Blot | |
| GABA(A) receptor subunit alpha-5, gamma-aminobutyric acid (GABA) A receptor, alpha 5, gamma-aminobutyric acid receptor subunit alpha-5, MGC138184 | |
| Mouse | |
| Affinity Purified | |
| Cellular Markers, Neuronal Cell Markers, Neurotransmission | |
| 2558.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction