missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA Receptor Epsilon Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97158AF594
This item is not returnable.
View return policy
Description
GABA Receptor Epsilon Polyclonal antibody specifically detects GABA Receptor Epsilon in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GABA Receptor Epsilon | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| GABA(A) receptor subunit epsilon, gamma-aminobutyric acid (GABA) A receptor, epsilon, gamma-aminobutyric acid receptor subunit epsilon | |
| Produced in rabbits immunized with E. coli-derived Human GABA Receptor Epsilon fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 2564 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction