missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA Transporter 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-1872
This item is not returnable.
View return policy
Description
GABA Transporter 2 Polyclonal specifically detects GABA Transporter 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| GABA Transporter 2 | |
| Polyclonal | |
| Unconjugated | |
| Antiserum with No Preservative | |
| GABA transport protein, GAT-2, GAT2Solute carrier family 6 member 13, sodium- and chloride-dependent GABA transporter 2, solute carrier family 6 (neurotransmitter transporter, GABA), member 13 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Reacts specifically with protein derived from CNS samples. | |
| Human, Mouse, Rat | |
| Antisera |
| Western Blot, Flow Cytometry | |
| LYOPH | |
| Western Blot 1/200-1/5000, Flow Cytometry | |
| P31646 | |
| SLC6A13 | |
| A 15 residue C-teminal domain synthetic peptide (Rat). | |
| 0.2 mL | |
| Neuroscience | |
| 6540 | |
| Reconstitute with 0.2 ml distilled water. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction