missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABARAP Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-158JF549
This item is not returnable.
View return policy
Description
GABARAP Polyclonal antibody specifically detects GABARAP in Human, Rat samples. It is validated for Western Blot
Specifications
| GABARAP | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| Janelia Fluor 549 | |
| ATG8A, FLC3B, FLJ25768, GABA(A) receptor-associated proteinMGC120154, gamma-aminobutyric acid receptor-associated protein, MM46MGC120155 | |
| N-terminus of GABAa Receptor Associated Protein (Proprietary) | |
| 0.1 mL | |
| Autophagy, Golgi Apparatus Markers, Macroautophagy, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 11337 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction