Learn More
Invitrogen™ GABARAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA14147
Descripción
PA1-4147 detects GABA Receptor Associated Protein in human, rat, mouse, bovine, and non-human primate samples. PA1-4147 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~116 kDa protein representing GABA Receptor Associated Protein in brain lysate. The PA1-4147 immunogen is a synthetic peptide corresponding to residues P(33) H L P R P H P R V P P H P S S E R R A V Y(54) of the mouse and rat GABA R1 subunit.
Gamma-aminobutyric acid (GABA) is the main inhibitory transmitter by increasing a Cl-conductance that inhibits neuronal firing in the central nervous system. It has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. GABARAP (GABAA receptor-associated protein) links GABAA receptors to the cytoskeleton and may play a role in intracellular transport of GABAA receptors and its interaction with the cytoskeleton. GABARAP belongs to the MAP1 or ATG8 like family and recent studies show that MAPK15/ERK8 is acting through interaction with ATG8 family proteins to regulate autophagy.
Especificaciones
| GABARAP | |
| Polyclonal | |
| Unconjugated | |
| GABARAP | |
| ATG8A; FLC3B; FLJ25768; GABA type A receptor-associated protein; GABA(A) receptor-associated protein; GABA-A receptor-associated protein; Gabarap; GABARAP a; GABARAP-a; gamma-aminobutyric acid receptor associated protein; gamma-aminobutyric acid receptor-associated protein; gamma-aminobutyric acid reseptor associated protein; HT004; hypothetical protein; MGC120154; MGC120155; MM46 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11337, 327715, 56486, 58974 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O95166, P60517, Q9DCD6, Q9GJW7 | |
| GABARAP | |
| Synthetic peptide corresponding to residues P(33) H L P R P H P R V P P H P S S E R R A V Y(54) of the mouse and rat GABA R1 subunit. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey, Bovine | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.