missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GABARAPA Polyclonal Antibody
GREENER_CHOICE

Product Code. 15960905
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15960905 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15960905 Supplier Invitrogen™ Supplier No. PA572235

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GABARAPA Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen GABARAPA
Applications Western Blot
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene gabarapa
Gene Accession No. 0
Gene Alias GABA(A) receptor associated protein; GABA(A) receptor-associated protein a; gabarap; gabarap protein; gabarapa; mg:bb02b03
Gene Symbols gabarapa
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 33-66 amino acids from the human region of DANRE gabarapa.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 326974
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.