missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABARAPL3 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18246IR
This item is not returnable.
View return policy
Description
GABARAPL3 Polyclonal antibody specifically detects GABARAPL3 in Human, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| GABARAPL3 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| ATG8D, GABARAPL3, Gamma-aminobutyric acid receptor-associated protein-like 3 | |
| Synthetic peptide from the C-terminal of Human GABARAPL3 | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 23766 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction