Learn More
Invitrogen™ GABBR1 Monoclonal Antibody (N93A/49), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545539
Description
1 μg/mL of MA5-45539 was sufficient for detection of GABA(B)R1 in 20 μg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 115kDa. No cross-reactivity against GABA(B)R2. This antibody was formerly sold as clone S93A-49.
Receptor for GABA. The activity of this receptor is mediated by G-proteins that inhibit adenylyl cyclase activity, stimulates phospholipase A2, activates potassium channels, inactivates voltage-dependent calcium-channels and modulates inositol phospholipids hydrolysis. Plays a critical role in the fine-tuning of inhibitory synaptic transmission. Pre-synaptic GABA-B-R inhibit neurotransmitter release by down-regulating high-voltage activated calcium channels, whereas postsynaptic GABA-B-R decrease neuronal excitability by activating a prominent inwardly rectifying potassium (Kir) conductance that underlies the late inhibitory postsynaptic potentials. Not only implicated in synaptic inhibition but also in hippocampal long-term potentiation, slow wave sleep, muscle relaxation and antinociception.
Specifications
| GABBR1 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Q9UBS5, Q9WV18, Q9Z0U4 | |
| GABBR1 | |
| Fusion protein amino acids 873-977 (cytoplasmic C-terminus) of rat GABA(B)R1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| N93A/49 | |
| PE | |
| GABBR1 | |
| bM573K1.1; bM573K1.1.1 (gamma-aminobutyric acid (GABA) B receptor, 1a); DAAP-188P13.3; dJ271M21.1.1; dJ271M21.1.2; FLJ92613; GABA(B) receptor; GABAB; GABA-B receptor 1; GABA-B receptor, R1 subunit; GABAB1; Gaba-b1; GABAbR1; GABA-BR1; GABA-B-R1; Gababr1 receptor; Gaba-b-r1a; Gaba-br1a receptor; GABA-BR1b receptor; Gabbr1; GABBR1-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor 1; gamma-aminobutyric acid (GABA) B receptor, 1; gamma-aminobutyric acid (GABA-B) receptor, 1; gamma-aminobutyric acid type B receptor subunit 1; GB1; GBR1; GPRC3A; HGB1a; OTTHUMP00000109099; OTTHUMP00000214725; seven transmembrane helix receptor | |
| Mouse | |
| Protein G | |
| RUO | |
| 2550, 54393, 81657 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.