missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABPB1 Mouse anti-Human, Clone: OTI4H6, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811263
This item is not returnable.
View return policy
Description
Transcription factor capable of interacting with purine rich repeats (GA repeats). Necessary for the expression of the Adenovirus E4 gene.Specifications
| GABPB1 | |
| Monoclonal | |
| Unconjugated | |
| GABPB1 | |
| BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4TF1B, GABPB, GABPB-1, GABPB2, NRF2B1, NRF2B2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2553 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI4H6 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q06547 | |
| GABPB1 | |
| Human recombit protein fragment corresponding to amino acids 128-360 of human GABPB1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction