Learn More
Invitrogen™ GABRA2 Monoclonal Antibody (N399/19), PerCP

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545727
Description
A 1:100 dilution of MA5-45727 was sufficient for detection of GABA-A R, Alpha2 in 20 μg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 55kDa. Does not cross-react with GABA-A Receptor Alpha1. This antibody was formerly sold as clone S399-19.
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| GABRA2 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| P23576, P26048, P47869 | |
| GABRA2 | |
| Fusion protein amino acids 350-385 (Cytoplasmic C-terminus) of rat GABA-A Receptor Alpha2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| N399/19 | |
| PerCP | |
| GABRA2 | |
| C630048P16Rik; FLJ97076; GABA(A) receptor; GABA(A) receptor subunit alpha-2; GABA(A) receptor, alpha 2; GABAARalpha2; Gabra2; Gabra-2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 2; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 2; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 2; gamma-aminobutyric acid A receptor alpha 2; gamma-aminobutyric acid receptor A2 subunit; gamma-aminobutyric acid receptor subunit alpha-2; gamma-aminobutyric acid type A receptor alpha2 subunit | |
| Mouse | |
| Protein G | |
| RUO | |
| 14395, 2555, 289606 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.