missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABRA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109750
This item is not returnable.
View return policy
Description
Immunogen sequence: ARNSLPKVAY ATAMDWFIAV CYAFVFSALI EFATVNYFTK RGWAWDGKSV VNDKKKEKAS VMIQNNAYAV AVANYAPNLS KDPVLSTISK SATTPEPNKK P.
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| GABRA2 | |
| Polyclonal | |
| Unconjugated | |
| GABRA2 | |
| C630048P16Rik; FLJ97076; GABA(A) receptor; GABA(A) receptor subunit alpha-2; GABA(A) receptor, alpha 2; GABAARalpha2; Gabra2; Gabra-2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 2; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 2; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 2; gamma-aminobutyric acid A receptor alpha 2; gamma-aminobutyric acid receptor A2 subunit; gamma-aminobutyric acid receptor subunit alpha-2; gamma-aminobutyric acid type A receptor alpha2 subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14395, 289606 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.67 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P23576, P26048 | |
| GABRA2 | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human GABRA2 (NP_000798.2). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction