missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GABRA3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16566501
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16566501 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16566501 Supplier Invitrogen™ Supplier No. PA588192

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QGESRRQEPG DFVKQDIGGL SPKHAPDIPD DSTDNITIFT RILDRLLDGY DNRLRPGLGD AVTEVKTDIY VTSFGPVSDT DMEYTIDVFF RQTWHDERLK FDGPMKILPL NNLLASKIWT PDTFFHNGKK SVAHNMTTPN KLLRLVDNGT LLYTMRLTIH AECPMHLEDF PMDVHACPLK FGSYAYTTAE VVYSWTLGKN KSVEVAQDGS RLNQYDLLGH VVGTEIIRSS TGEYVVMTTH FHLKRKIG; Positive Samples: SH-SY5Y, Mouse kidney, Rat liver; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse.

GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRA3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Gabra3
Gene Accession No. P20236, P26049, P34903
Gene Alias GABA(A) receptor; GABA(A) receptor subunit alpha-3; GABA(A) receptor, alpha 3; GABA-A receptor alpha-3 subunit; Gabra3; Gabra-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 3; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 3; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 3; gamma-aminobutyric acid receptor subunit alpha-3; gamma-aminobutyric acid receptor, subunit alpha 3; gamma-aminobutyric acid type A receptor alpha3 subunit; gamma-aminobutyric acid type A receptor subunit alpha3; Gamma-animobutyric acid (GABA) A receptor alpha 3; gamma-animobutyric acid (GABA) A receptor, alpha 3; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit alpha-3; gamma-aminobutyric acid type A receptor subunit alpha3; MGC33793; unnamed protein product
Gene Symbols Gabra3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 29-276 of human GABRA3 (NP_0007991).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14396, 24947, 2556
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.