Learn More
GABRA5 Monoclonal Antibody (OTI1G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806598
This item is not returnable.
View return policy
Description
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Transcript variants utilizing three different alternative non-coding first exons have been described.Specifications
| GABRA5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P31644 | |
| GABRA5 | |
| Human recombit protein fragment corresponding to amino acids 32-259 of human GABRA5 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1G5 | |
| Unconjugated | |
| GABRA5 | |
| gamma-aminobutyric acid type A receptor alpha5 subunit | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2558 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |