missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRA5 Monoclonal Antibody (OTI1G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806473S
This item is not returnable.
View return policy
Description
GABRA5 Monoclonal antibody specifically detects GABRA5 in Human samples. It is validated for Western BlotSpecifications
| GABRA5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13772 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8031 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1G9 | |
| Unconjugated | |
| NCOA4 | |
| A230018I05Rik; GABA(A) receptor; GABA(A) receptor subunit alpha-5; GABAA receptor alpha5 subunit; GABA-A receptor alpha-5 subunit; GABRA5; Gabra-5; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, alpha 5; gamma-aminobutyric acid (GABA) A receptor, alpha 5; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 5; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 5; gamma-aminobutyric acid A receptor, alpha 5; gamma-aminobutyric acid receptor subunit alpha-5; gamma-aminobutyric acid type A receptor alpha5 subunit; ligand-gated ion channel subunit | |
| Human recombinant protein fragment corresponding to amino acids 280-553 of human NCOA4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction