missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABRB1 Recombinant Rabbit Monoclonal Antibody (1Q3A9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537949
This item is not returnable.
View return policy
Description
Positive Samples: SH-SY5Y, Mouse spinal cord, Mouse brain, Rat brain Immunogen sequence: EKNKLEMNKV QVDAHGNILL STLEIRNETS GSEVLTSVSD PKATMYSYDS ASIQYRKPLS SREAYGRALD RHGVPSKGRI RRRASQLKVK IPDLTDVNSI D.
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the GABA/benzodiazepine receptor and opening an integral chloride channel.
Specifications
| GABRB1 | |
| Recombinant Monoclonal | |
| 0.3 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15431, P18505, P50571 | |
| Gabrb1 | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human GABA A Receptor beta 1 (GABRB1) (NP_0008032). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1Q3A9 | |
| Unconjugated | |
| Gabrb1 | |
| AW061132; B230208N19Rik; GABA beta subunit; GABA(A) receptor; GABA(A) receptor subunit beta-1; GABA(A) receptor, beta 1; GABA(A)R beta-1; GABA-RB; Gabrb1; Gabrb-1; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 1; gamma-aminobutyric acid (GABA) A receptor, subunit beta 1; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 1; gamma-aminobutyric acid A receptor beta 1; Gamma-aminobutyric acid receptor beta 1; gamma-aminobutyric acid receptor subunit beta-1; gamma-aminobutyric acid receptor, subunit beta 1; gamma-aminobutyric acid type A receptor beta1 subunit; gamma-aminobutyric acid type A receptor subunit beta1; GARB1; GBRB1; si:dkey-20i10.4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14400, 25450, 2560 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction