missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAD1/GAD67 Antibody (OTI3G9), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70791H
This item is not returnable.
View return policy
Description
GAD1/GAD67 Monoclonal specifically detects GAD1/GAD67 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| GAD1/GAD67 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| GAD1 | |
| Full length recombinant protein of human GAD1/GAD67 (NP_000808) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI3G9 | |
| Western Blot, Immunohistochemistry | |
| 67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform | |
| Mouse | |
| Affinity Purified | |
| Diabetes Research | |
| 2571.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction