missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAD2/GAD65 Antibody (5D9G12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 568.00
Specifications
| Antigen | GAD2/GAD65 |
|---|---|
| Clone | 5D9G12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
Description
GAD2/GAD65 Monoclonal specifically detects GAD2/GAD65 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| GAD2/GAD65 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience | |
| PBS with 0.05% Sodium Azide | |
| 2572 | |
| Purified recombinant fragment of human GAD2 (AA: 1-100) expressed in E. Coli. | |
| Primary | |
| 65.4 kDa |
| 5D9G12 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 4.1.1, EC 4.1.1.15, GAD-65, GAD65MGC161605, glutamate decarboxylase 2, glutamate decarboxylase 2 (pancreatic islets and brain, 65kD), glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa), Glutamate decarboxylase 65 kDa isoform, Glutamate decarboxylase-2 (pancreas), MGC161607,65 kDa glutamic acid decarboxylase | |
| GAD2 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title