missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAD2/GAD65 Antibody (GAD2/8394R), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21107C
This item is not returnable.
View return policy
Description
GAD2/GAD65 Monoclonal antibody specifically detects GAD2/GAD65 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GAD2/GAD65 | |
| Monoclonal | |
| DyLight 650 | |
| EC 4.1.1, EC 4.1.1.15, GAD-65, GAD65MGC161605, glutamate decarboxylase 2, glutamate decarboxylase 2 (pancreatic islets and brain, 65kD), glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa), Glutamate decarboxylase 65 kDa isoform, Glutamate decarboxylase-2 (pancreas), MGC161607,65 kDa glutamic acid decarboxylase | |
| Recombinant human GAD2/GAD65 protein fragment (around aa 1-200) (exact sequence is proprietary) | |
| 0.1 mL | |
| Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience | |
| 2572 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| GAD2/8394R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction