missing translation for 'onlineSavingsMsg'
Learn More

GAD2 / GAD65 (GABAergic Neuronal Marker) Mouse, Clone: GAD2/1960, enQuireBio™

Mouse Monoclonal Antibody

€ 199.00 - € 510.00

Specifications

Antigen GAD2 / GAD65 (GABAergic Neuronal Marker)
Clone GAD2/1960
Concentration 200μg/mL
Applications ELISA, Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15940078
View Documents Promotion Details
EnquireBio
2572-MSM1-P1
100 μg
€ 510.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15930078
View Documents Promotion Details
EnquireBio
2572-MSM1-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

GAD2 / GAD65 (GABAergic Neuronal Marker)
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant human GAD2 (GAD65) protein fragment (apprx. aa 6-99)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
65kDa
GAD2/1960
ELISA, Western Blot, Immunohistochemistry
Unconjugated
RUO
2572
IgG2b κ
Protein A/G purified from Bioreactor concentrate.
Cytoplasmic
Pancreas or Brain.
Videos
SDS
Documents

Documents

Product Certifications