missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GADD34 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10449-1-AP-20UL
This item is not returnable.
View return policy
Description
Molecular weight of GADD34 is 100 kDa confirmed in GADD34 KO mice, and Proteintech's GADD34 antibody 10449-1-AP primarily recognize the 100 kDa band.
This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The induction of this gene by ionizing radiation occurs in certain cell lines regardless of p53 status, and its protein response is correlated with apoptosis following ionizing radiation.Specifications
| GADD34 | |
| Polyclonal | |
| Unconjugated | |
| PPP1R15A | |
| GADD34, growth arrest and DNA damage-inducible protein 34, PPP1R15A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171071, 17872, 23645 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75807, P17564, Q6IN02 | |
| Ppp1r15a | |
| GADD34 Fusion Protein Ag0578 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction