missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GADD45A Monoclonal Antibody (OTI1C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507370S
This item is not returnable.
View return policy
Description
GADD45A Monoclonal antibody specifically detects GADD45A in Human samples. It is validated for Western BlotSpecifications
| GADD45A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NQG5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 58490 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1C9 | |
| Unconjugated | |
| RPRD1B | |
| AA545191; DDIT 1; DDIT1; DDIT-1; DNA damage-inducible transcript 1 protein; DNA damage-inducible transcript-1; DNA-damage inducible transcript 1; DNA-damage-inducible transcript 1; GADD45; GADD45A; Gadd45alpha; growth arrest and DNA damage inducible alpha; growth arrest and DNA damage-inducible protein GADD45 alpha; growth arrest and DNA-damage-inducible 45 alpha; growth arrest and DNA-damage-inducible protein GADD45 alpha; growth arrest and DNA-damage-inducible, alpha | |
| Full length human recombinant protein of human RPRD1B produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction