Learn More
GADD45G Monoclonal Antibody (OTI5G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505535
Description
GADD45G gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The protein encoded by this gene responds to environmental stresses by mediating activation of the p38/JNK pathway via MTK1/MEKK4 kinase. The GADD45G is highly expressed in placenta.Specifications
| GADD45G | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95257 | |
| GADD45G | |
| Full length human recombit protein of human GADD45G produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI5G2 | |
| Unconjugated | |
| GADD45G | |
| CR6, DDIT2, GADD45gamma, GRP17 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10912 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.