missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GADD45G Monoclonal Antibody (OTI7A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505534
This item is not returnable.
View return policy
Description
GADD45G gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The protein encoded by this gene responds to environmental stresses by mediating activation of the p38/JNK pathway via MTK1/MEKK4 kinase. The GADD45G is highly expressed in placenta.Specifications
| GADD45G | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95257 | |
| GADD45G | |
| Full length human recombit protein of human GADD45G produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI7A3 | |
| Unconjugated | |
| GADD45G | |
| CR6, DDIT2, GADD45gamma, GRP17 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10912 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction