missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gag polyprotein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 299.00 - € 507.00
Specifications
| Antigen | Gag polyprotein |
|---|---|
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614699
|
Novus Biologicals
NBP3-12744-50ul |
50 μg |
€ 299.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657668
|
Novus Biologicals
NBP3-12744-100ul |
100 μg |
€ 507.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gag polyprotein Polyclonal antibody specifically detects Gag polyprotein in Virus samples. It is validated for Western Blot, ELISASpecifications
| Gag polyprotein | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Pr55Gag | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant Gag polyprotein (Accession#: NP_057849.4; Pro588-Phe1027). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title