missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAK Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32181AF647
This item is not returnable.
View return policy
Description
GAK Polyclonal antibody specifically detects GAK in Human, Mouse samples. It is validated for Western Blot
Specifications
| GAK | |
| Polyclonal | |
| Western Blot | |
| cyclin G associated kinase, cyclin-G-associated kinase, EC 2.7.11.1, FLJ16629, FLJ40395, MGC99654 | |
| The immunogen this antibody was made to, maps to a region between residue 675 to 725 of human Cyclin G Associated Kinase using the numbering given in entry NP_005246.2 (GeneID 2580). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 2580 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction