Learn More
Invitrogen™ GAL3ST1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551541
Description
Immunogen sequence: LLNILFRFGQ KHRLKFAFPN GRNDFDYPTF FARSLVQDYR PGACFNIICN HMRFHYDEVR GLVPTNAIFI TVLRDPARLF ESSFHYFGPV VPLTWKLSAG DKLTEFLQDP DRYYDPNGFN AHYLRNLLFF DLGYDNSLDP SSPQVQEHIL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
Catalyzes the sulfation of membrane glycolipids. Seems to prefer beta-glycosides at the non-reducing termini of sugar chains attached to a lipid moiety. Catalyzes the synthesis of galactosylceramide sulfate (sulfatide), a major lipid component of the myelin sheath and of monogalactosylalkylacylglycerol sulfate (seminolipid), present in spermatocytes. Also acts on lactosylceramide, galactosyl 1-alkyl-2-sn-glycerol and galactosyl diacylglycerol (in vitro).
Specifications
| GAL3ST1 | |
| Polyclonal | |
| Unconjugated | |
| Gal3st1 | |
| 3'-phosphoadenosine-5'-phosphosulfate:GalCer sulfotransferase; 3'-phosphoadenylylsulfate:galactosylceramide 3'-sulfotransferase; 3'-phosphoadenylylsulfate-galactosylceramide 3'-sulfotransferase; cerebroside (3'-phosphoadenylylsulfate:galactosylceramide 3') sulfotransferase; cerebroside sulfotransferase; Cst; Gal3st1; galactose-3-O-sulfotransferase 1; galactosylceramide sulfotransferase; GalCer sulfotransferase; Gcst | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9514 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99999 | |
| Gal3st1 | |
| Recombinant protein corresponding to Human GAL3ST1. Recombinant protein control fragment (Product #RP-91367). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.