missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galanin Antibody (581403), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5854S
This item is not returnable.
View return policy
Description
Galanin Monoclonal antibody specifically detects Galanin in Human samples. It is validated for Immunohistochemistry, Flow Cytometry, CyTOF-reported
Specifications
| Galanin | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Flow Cytometry, CyTOF | |
| 581403 | |
| Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-reported | |
| GAL1, galanin, galanin prepropeptide, GALNgalanin-related peptide, GLNNgalanin-message-associated peptide, GMAP, MGC40167 | |
| E.coli-derived recombinant human Galanin, Ala20-Ser123, Accession # P22466 | |
| 0.1 mL | |
| Neuroscience | |
| 51083 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction