missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galanin Antibody (581403), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5854M
This item is not returnable.
View return policy
Description
Galanin Monoclonal antibody specifically detects Galanin in Human samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF-reported
Specifications
| Galanin | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 581403 | |
| Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-reported | |
| GAL1, galanin, galanin prepropeptide, GALNgalanin-related peptide, GLNNgalanin-message-associated peptide, GMAP, MGC40167 | |
| E.coli-derived recombinant human Galanin, Ala20-Ser123, Accession # P22466 | |
| 0.1 mL | |
| Neuroscience | |
| 51083 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction