Learn More
Galanin Antibody (581403), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB5854Y
Product Code. 18520377
  • € 706.00 / 0.10mL
Description
Galanin Monoclonal antibody specifically detects Galanin in Human samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF-reported
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Galanin
Monoclonal
DyLight 680
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
581403
Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-reported
GAL1, galanin, galanin prepropeptide, GALNgalanin-related peptide, GLNNgalanin-message-associated peptide, GMAP, MGC40167
E.coli-derived recombinant human Galanin, Ala20-Ser123, Accession # P22466
0.1 mL
Neuroscience
51083
Store at 4°C in the dark.
IgG2b