missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Galanin Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Specifications
Specifications
| Gene ID (Entrez) | 51083 |
| Species | Human |
| Content And Storage | Store at -80C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP1-45217. For further blocking peptide related protocol, click here . |
| Gene Alias | GAL1, galanin, galanin prepropeptide, GALNgalanin-related peptide, GLNNgalanin-message-associated peptide, GMAP, MGC40167 |
| Quantity | 0.1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction