missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galanin R2/GALR2 Antibody (616520) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6544F
This item is not returnable.
View return policy
Description
Galanin R2/GALR2 Monoclonal antibody specifically detects Galanin R2/GALR2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| Galanin R2/GALR2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 616520 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| GAL2-R, galanin receptor 2, galanin receptor type 2, GALNR2MGC125984, GALR-2, MGC125983 | |
| NS0 mouse myeloma cell line transfected with human Galanin R2, Accession # O43603 | |
| 0.1 mL | |
| GPCR | |
| 8811 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction