missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galanin R2/GALR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00
Specifications
| Antigen | Galanin R2/GALR2 |
|---|---|
| Dilution | Western Blot 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Galanin R2/GALR2 Polyclonal specifically detects Galanin R2/GALR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Galanin R2/GALR2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| GAL2-R, galanin receptor 2, galanin receptor type 2, GALNR2MGC125984, GALR-2, MGC125983 | |
| GALR2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| GPCR | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 8811 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RGTHSGSVLERESSDLLHMSEAAGALRPCPGASQPCILEPCPGPSWQGPKAGDSILTVDVA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title