missing translation for 'onlineSavingsMsg'
Learn More

Galanthaminone, MedChemExpress

Product Code. p-200101249
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16443826

Brand: Medchem Express HYI0020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Galanthaminone (Narwedin) is a competitive and reversible cholinesterase (AChE) inhibitor; is used for the treatment of mild to moderate Alzheimer's disease and various other memory impairments.

Galanthaminone (Narwedin) is a competitive and reversible cholinesterase (AChE) inhibitor

Specifications

Chemical Name or Material Galanthaminone
CAS 510-77-0
Color White
Purity Grade Notes Research
Molecular Formula C17H19NO3
Quantity 25 mg
Synonym (-)-Narwedine Narwedin
Solubility Information DMSO : 10 mg/mL (35.05 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C=C[C@@]23CCN(C)CC4=CC=C(OC)C(O[C@@]3([H])C1)=C24
Molecular Weight (g/mol) 285.34
Formula Weight 285.34
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.