missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-1 Antibody (RM0081-9J26), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-21721G
This item is not returnable.
View return policy
Description
Galectin-1 Monoclonal specifically detects Galectin-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Galectin-1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LGALS1 | |
| Mouse recombinant Galectin-1 | |
| 0.1 mL | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Galectin-1. | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| RM0081-9J26 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
| 14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1 | |
| Rat | |
| Protein A or G purified | |
| Apoptosis, Biologically Active Proteins | |
| 3956 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction