missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galectin 12 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145433
This item is not returnable.
View return policy
Description
Galectin-12, also designated galectin-related inhibitor of proliferation 1 in mouse, is a 314 amino acid protein encoded by the human gene LGALS12. Galectin-12 is a member of the galectin family consisting of b-galactoside-binding proteins with conserved carbohydrate recognition domains. Galectin-12 binds lactose and may participate in the apoptosis of adipocytes. This protein is preferentially expressed in peripheral blood leukocytes and adipocytes. Galectin-12 is induced by cell cycle block at the G1 phase and causes G1 arrest when overexpressed. The galectin-12 gene is expressed in mouse preadipo-cytes and is upregulated when preadipocytes undergo cell cycle arrest, concomitant with acquisition of the competence to undergo differentiation in response to adipogenic hormone stimulation. Galectin-12 is an adipocyte-expressed protein which is downregulated by various insulin resistance-inducing hormones. As a result, galectin-12 may play a role in the pathogenesis of insulin resistance.
Specifications
| Galectin 12 | |
| Polyclonal | |
| Unconjugated | |
| LGALS12 | |
| LGALS12 | |
| Recombinant Pig GAL12 protein, Met1-Tyr314 (Accession #B7U2G9). | |
| 200 μL | |
| Primary | |
| Pig | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| AW987437; GAL12; gal-12; galectin 12; galectin-12; Galectin-related inhibitor of proliferation; galectin-related inhibitor of proliferation 1; GRIP1; lectin, galactose binding, soluble 12; lectin, galactoside binding soluble 12; lectin, galactoside-binding, soluble, 12; LGALS12; testicular secretory protein Li 26 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100217395 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction