missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-14/LGALS14 Antibody (S09-6C1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19676-100ul
This item is not returnable.
View return policy
Description
Galectin-14/LGALS14 Monoclonal antibody specifically detects Galectin-14/LGALS14 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Galectin-14/LGALS14 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 56891 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S09-6C1 | |
| Unconjugated | |
| Charcot-Leyden crystal protein 2, CLC2, gal-14, galectin-14, lectin, galactoside-binding, soluble, 14, MGC22235, Placental Protein 13-Like Protein, PPL13 | |
| A synthetic peptide of human Galectin-14/LGALS14 (Uniprot # Q8TCE9) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction