missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-2 Antibody (2509A) [Janelia Fluor™ 549], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB1153I
This item is not returnable.
View return policy
Description
Galectin-2 Monoclonal antibody specifically detects Galectin-2 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| Galectin-2 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 2509A | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Beta-galactoside-binding lectin L-14-II, Gal-2, galectin 2, galectin-2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, MGC75071, S-Lac lectin 2 | |
| E. coli-derived human Galectin-2, Met1-Glu132, Accession # P05162 | |
| 0.1 mL | |
| Apoptosis, Cancer, Cellular Markers, Tumor Suppressors | |
| 3957 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction