missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-2 Antibody (OTI6A8), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71123AF700
This item is not returnable.
View return policy
Description
Galectin-2 Monoclonal specifically detects Galectin-2 in Human samples. It is validated for Western Blot.
Specifications
| Galectin-2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LGALS2 | |
| Full length recombinant protein of human Galectin-2 (NP_006489) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI6A8 | |
| Western Blot | |
| Beta-galactoside-binding lectin L-14-II, Gal-2, galectin 2, galectin-2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, MGC75071, S-Lac lectin 2 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer, Cellular Markers, Tumor Suppressors | |
| 3957.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction