Learn More
Galectin-3 Antibody (194801), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB11542MFV450
Product Code. 18508995
  • € 674.00 / 0.10mL
Description
Galectin-3 Monoclonal antibody specifically detects Galectin-3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Galectin-3
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
0.1 mL
Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Neuroscience, Tumor Suppressors
3958
Store at 4C in the dark.
Western Blot, Flow Cytometry, CyTOF
194801
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3
E. coli-derived recombinant human Galectin-3, Ala2-Ile250, Accession # P17931
RUO
Primary
Human
IgG2b