missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-3 Antibody (5C21), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-92690H
This item is not returnable.
View return policy
Description
Galectin-3 Monoclonal specifically detects Galectin-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Galectin-3 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| LGALS3 | |
| Full length recombinant human Galectin-3 expressed in and purified from E. coli. [UniProt# P17931] | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Neuroscience, Tumor Suppressors | |
| 3958 | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry | |
| 5C21 | |
| Western Blot, Immunohistochemistry | |
| Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction