Learn More
Galectin-3 Antibody (OTI1C7), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71124JF549
Code produit. 18172973
  • € 751.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Galectin-3 Monoclonal specifically detects Galectin-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Galectin-3
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
LGALS3
Full length recombinant protein of human Galectin-3 (NP_002297) produced in E.coli.
0.1 mL
Primary
Human, Mouse
IgG
Western Blot, Immunohistochemistry
OTI1C7
Western Blot, Immunohistochemistry
Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3
Mouse
Affinity Purified
Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Neuroscience, Tumor Suppressors
3958.0
Store at 4°C in the dark.