missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-3 Antibody (S05-8J4), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 421.00 - € 645.00
Specifications
| Antigen | Galectin-3 |
|---|---|
| Clone | S05-8J4 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18676514
|
Novus Biologicals
NBP3-20038-100ul |
100 μg |
€ 645.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665824
|
Novus Biologicals
NBP3-20038-50ul |
50 μg |
€ 421.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Galectin-3 Monoclonal antibody specifically detects Galectin-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Galectin-3 | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Neuroscience, Tumor Suppressors | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 3958 | |
| IgG | |
| Affinity purified |
| S05-8J4 | |
| Western Blot, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3 | |
| A synthetic peptide of human Galectin-3 (Uniprot # P17931) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title