Learn More
Galectin-3BP/MAC-2BP/LGALS3BP Antibody (OTI6B7), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71125PEATT594
Product Code. 18110334
  • € 839.00 / 0.10mL
Description
Galectin-3BP/MAC-2BP/LGALS3BP Monoclonal specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Galectin-3BP/MAC-2BP/LGALS3BP
Monoclonal
PE/ATTO 594
PBS with 0.05% Sodium Azide
LGALS3BP
Recombinant protein fragment corresponding to amino acids 19-300 of human Galectin-3BP/MAC-2BP/LGALS3BP (NP_005558) produced in E.coli.
0.1 mL
3959.0
Store at 4°C in the dark. Do not freeze.
Flow Cytometry
OTI6B7
Flow Cytometry
90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K
Mouse
Affinity Purified
Primary
Human
IgG2a