missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galectin 4 Recombinant Rabbit Monoclonal Antibody (9Z2B5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538087
This item is not returnable.
View return policy
Description
Positive Samples: HT-29, Rat large intestine Immunogen sequence: MAYVPAPGYQ PTYNPTLPYY QPIPGGLNVG MSVYIQGVAS EHMKRFFVNF VVGQDPGSDV AFHFNPRFDG WDKVVFNTLQ GGKWGSEERK RSMPFKKGAA.
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The expression of this gene is restricted to small intestine, colon, and rectum, and it is underexpressed in colorectal cancer. Galectin 4 binds lactose and a related range of sugars. May be involved in the assembly of adherens junctions.
Specifications
| Galectin 4 | |
| Recombinant Monoclonal | |
| 0.11 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 | |
| P38552, P56470, Q8K419 | |
| Lgals4 | |
| Synthetic Peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 9Z2B5 | |
| Unconjugated | |
| Lgals4 | |
| Antigen NY-CO-27; GAL4; gal-4; galectin 4; galectin-4; L-36; L-36 lactose-binding protein; L36LBI; L36LBP; lactose-binding lectin 4; Lectin galactose binding soluble 4 (Galectin-4); lectin, galactose binding, soluble 4; Lectin, galactose binding, soluble 4 (Galectin-4); lectin, galactoside binding soluble 4; lectin, galactoside-binding, soluble, 4; lectin, galactoside-binding, soluble, 4 (galectin 4); Lgals4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16855, 25474, 3960 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction