missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-7 Antibody (950723) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB13392F
This item is not returnable.
View return policy
Description
Galectin-7 Monoclonal antibody specifically detects Galectin-7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Specifications
| Galectin-7 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 950723 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready | |
| Gal-7, GAL7, galectin 7, galectin-7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1 | |
| E. coli-derived recombinant human Galectin-7, Ser2-Phe136, Accession # NP_002298 | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 3963 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction